Learn More
TBC1D4, p Thr642 Antibody (SR1272), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22607-100ul
This item is not returnable.
View return policy
Description
TBC1D4 Monoclonal antibody specifically detects TBC1D4 in Human samples. It is validated for Western Blot
Specifications
| TBC1D4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1272 | |
| Western Blot 1:500-1:2000 | |
| Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4 | |
| A synthesized peptide derived from human phospho-TBC1D4 [Thr642] (Uniprot #: O60343) | |
| 100 μg | |
| Apoptosis, B Cell Development and Differentiation Markers, Diabetes Research, Lipid and Metabolism, Phospho Specific, Signal Transduction | |
| 9882 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |