missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557042
This item is not returnable.
View return policy
Description
Immunogen sequence: TRRFVNQLNT KYRDSLPQLP KAFEQYLNLE DGRLLTHLRM CSAAPKLPYD LWFKRCFAGC LP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
TBC1D7 belongs to the family of proteins sharing a 180- to 200-amino acid TBC domain presumed to have a role in regulating cell growth and differentiation. These proteins share significant homology with TRE2.
Specifications
| TBC1D7 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D7 | |
| 2610009C09Rik; Cell migration-inducing protein 23; dJ257A7.3; HSPC239; MGCPH; PIG51; TBC1 domain family member 7; TBC1 domain family, member 7; TBC1D7; TBC7; TS complex subunit 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51256, 67046 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9D0K0, Q9P0N9 | |
| TBC1D7 | |
| Recombinant protein corresponding to Human TBC1D7. Recombinant protein control fragment (Product #RP-94877). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction