Learn More
Invitrogen™ TBC1D8B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555647
Description
Immunogen sequence: ASQDGNQCSV IIPLREVLAI DKTNDSSKSV IISIKGKTAF RFHEVKDFEQ LVAKLRLRCG AASTQYHDIS TELAISSEST EPSDNFEVQS LTSQRECSKT VNTEALMTVF HPQNLETLNS KMLKEKMKEQ SWKILFAECG RGVSMFRT Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
This gene encodes a protein with a TBC (Tre-2/Bub2/CDC16) domain. Some mammalian proteins with this domain have been shown to function as Rab-GAPs by binding to specific Rab proteins and affecting their GTPase activity. Alternative splicing results in multiple transcript variants.
Specifications
| TBC1D8B | |
| Polyclonal | |
| Unconjugated | |
| TBC1D8B | |
| 4921505D17Rik; 9030605E16Rik; GRAMD8B; RGD1563084; RP11-321G1.1; TBC1 domain family member 8B; TBC1 domain family, member 8B; TBC1 domain family, member 8B (with GRAM domain); TBC1D8B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54885 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q0IIM8 | |
| TBC1D8B | |
| Recombinant protein corresponding to Human TBC1D8B. Recombinant protein control fragment (Product #RP-94765). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.