missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D9B Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98499AF594
This item is not returnable.
View return policy
Description
TBC1D9B Polyclonal antibody specifically detects TBC1D9B in Human samples. It is validated for Western Blot
Specifications
| TBC1D9B | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 23061 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| KIAA0676FLJ41880, MGC157757, TBC1 domain family member 9B, TBC1 domain family, member 9B (with GRAM domain) | |
| Produced in rabbits immunized with E. coli-derived Human TBC1D9B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction