missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCA Antibody (1A5), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30168JF646
This item is not returnable.
View return policy
Description
TBCA Monoclonal antibody specifically detects TBCA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| TBCA | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CFA, chaperonin cofactor A, TCP1-chaperonin cofactor A, tubulin folding cofactor A, Tubulin-folding cofactor A, tubulin-specific chaperone a | |
| Recombinant TBCA (1-108aa) purified from E. coli | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 6902 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 1A5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction