missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TBCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30200
This item is not returnable.
View return policy
Description
TBCA Polyclonal antibody specifically detects TBCA in Human samples. It is validated for Western Blot
Specifications
| TBCA | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| CFA, chaperonin cofactor A, TCP1-chaperonin cofactor A, tubulin folding cofactor A, Tubulin-folding cofactor A, tubulin-specific chaperone a | |
| Between 58 and 108 | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 6902 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction