Learn More
Invitrogen™ TBCC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110460
Description
Immunogen sequence: MESVSCSAAA VRTGDMESQR DLSLVPERLQ RREQERQLEV ERRKQKRQNQ EVEKENSHFF VATFARERAA VEELLERAES VERLEEAASR LQGLQKLIND SVFFLAAYDL RQGQEALARL QAALAERRRG LQPKKRFAFK TRGKDAASST KVDAAPGIPP AVESIQDSPL PKKAEGDLGP SWVCGFSNLE SQVLEKRASE LHQRDVLLTE LSNCTVRLYG NPNTLRLTKA HSCKLLCGPV STSVFLEDCS DCVLAVACQQ LRIHSTKDTR IFLQVTSRAI VEDCSGIQFA PYTWSYPEID KDFESSGLDR SKNNWNDVDD FNWLARDMAS PNWSILPEEE RNIQWD.
Tubulin-folding protein; involved in the final step of the tubulin folding pathway.
Specifications
| TBCC | |
| Polyclonal | |
| Unconjugated | |
| TBCC | |
| 2810055C19Rik; CFC; EGK_14912; Tbcc; tubulin folding cofactor C; tubulin-folding cofactor C; Tubulin-specific chaperone C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 316221, 6903, 72726 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 3.3 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q15814, Q8VCN9 | |
| TBCC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-346 of human TBCC (NP_003183.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.