missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCCD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 232.00 - € 470.00
Specifications
| Antigen | TBCCD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617791
|
Novus Biologicals
NBP2-94513-0.02ml |
0.02 mL |
€ 232.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647510
|
Novus Biologicals
NBP2-94513-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBCCD1 Polyclonal antibody specifically detects TBCCD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TBCCD1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 55171 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ10560, TBCC domain containing 1, TBCC domain-containing protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 456-557 of human TBCCD1 (NP_060608.1). RVFQLLPPCEFYVFIIPFEMEGDTTEIPGGLPSVYQKALGQREQKIQIWQKTVKEAHLTKDQRKQFQVLVENKFYEWLINTGHRQQLDSLVPPAAGSKQAAG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto