missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TBCE Polyclonal Antibody

Product Code. 16606834
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16606834 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16606834 Supplier Invitrogen™ Supplier No. PA5100346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TBCE.

Cofactor E is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBCE
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TBCE
Gene Accession No. Q15813, Q5FVQ9, Q8CIV8
Gene Alias 2610206D02Rik; beta-tubulin cofactor E; C530005D02Rik; HRD; hypoparathyroidism, growth and mental retardation, and dysmorphism; hypothetical protein LOC664760; KCS; KCS1; pac2; pmn; Tbce; Tbce protein; tubulin folding cofactor E; tubulin-folding cofactor E; Tubulin-specific chaperone E; tubulin-specific chaperone E-like protein; Unknown (protein for MGC:133938); zgc:123075
Gene Symbols TBCE
Host Species Rabbit
Immunogen A synthesized peptide derived from human TBCE(Accession Q15813), corresponding to amino acid residues R24-K74.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361255, 6905, 70430
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.