missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TbHsp70 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18236JF585
This item is not returnable.
View return policy
Description
TbHsp70 Polyclonal antibody specifically detects TbHsp70 in Protozoa samples. It is validated for Western Blot
Specifications
| TbHsp70 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Protozoa | |
| Purified |
| Western Blot | |
| Janelia Fluor 585 | |
| heat shock 70 kDa protein 4 | |
| Synthetic peptide from the mid-protein of Trypanosoma brucei brucei TbHsp70 (aa. 200-300) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction