missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tbj51 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18235DL594
This item is not returnable.
View return policy
Description
Tbj51 Polyclonal antibody specifically detects Tbj51 in Protozoa samples. It is validated for Western Blot
Specifications
| Tbj51 | |
| Polyclonal | |
| Western Blot | |
| Tb04.1H19.310, TPR-repeat-containing chaperone protein DNAJ | |
| Synthetic peptide from the C-terminal of Trypanosoma brucei brucei Tbj51 (aa. 605-705) | |
| 0.1 mL | |
| Primary | |
| Protozoa | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 3656749 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction