missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBK1 Antibody (108A429), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56705R
This item is not returnable.
View return policy
Description
TBK1 Monoclonal specifically detects TBK1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| TBK1 | |
| Monoclonal | |
| DyLight 550 | |
| Q9UHD2 | |
| TBK1 | |
| A synthetic peptide corresponding to amino acids 563-577 (YYQFKKDKAERRLAYC) of human TBK1 was used as immunogen for this antibody (NP_037386). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross reacts with Cow, Dog, Human, Mouse and Rat. Predicted to react with Chimpanzee. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| 108A429 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated | |
| EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1 | |
| Mouse | |
| 84 kDa | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 29110 | |
| Human, Mouse, Rat, Bovine, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction