missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBK1 Antibody (14D10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26292-100ul
This item is not returnable.
View return policy
Description
TBK1 Monoclonal antibody specifically detects TBK1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| TBK1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 14D10 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1 | |
| A synthesized peptide derived from Human TBK1 [UniProt Q9UHD2] | |
| 100 μL | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 29110 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction