missing translation for 'onlineSavingsMsg'
Learn More

TBK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 227.00 - € 326.00

Specifications

Antigen TBK1
Dilution Immunohistochemistry-Paraffin 1:50-1:200
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18626052
View Documents
Novus Biologicals
NBP2-97957-100ul
100 μg
€ 326.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18695934
View Documents
Novus Biologicals
NBP2-97957-50ul
50 μg
€ 227.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TBK1 Polyclonal antibody specifically detects TBK1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TBK1
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Protein Kinase, Signal Transduction
PBS (pH 7.0)
29110
IgG
Antigen and protein A Affinity-purified
Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Purified
RUO
Human
EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1
Produced in rabbits immunized with E. coli-derived Human TBK1 fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.