missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBL1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB300-872
This item is not returnable.
View return policy
Description
TBL1 Polyclonal specifically detects TBL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| TBL1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| EBI, SMAP55, TBL1transducin (beta)-like 1, transducin (beta)-like 1X-linked, X-linked | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is also expected to recognize the human protein TBLR1 (NP_078941), which is very similar. Variants (NP_005638.1; NP_001132938.1) encode isoform a (TBL1X) and variants (NP_001132939.1; NP_001132940.1 ) encode isoform b (TBLX1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 4 - 6 ug/ml, Immunohistochemistry-Paraffin 4 - 6 ug/ml, Peptide ELISA Detection limit 1:4000 | |
| NP_005638. | |
| TBL1X | |
| Peptide with sequence SASDGSVCVLDLRK corresponding to C-Terminus according to NP_005638.1, NP_001132938.1, NP_001132939.1, NP_001132940.1, NP_078941.2. | |
| 0.1 mg | |
| Wnt Signaling Pathway | |
| 6907 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction