missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TBL1X/TBL1XR1 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA131502

Product Code. 11566123

  • € 578.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX22243-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation. This encoded protein is found as a subunit in corepressor SMRT (silencing mediator for retinoid and thyroid receptors) complex along with histone deacetylase 3 protein. This gene is located adjacent to the ocular albinism gene and it is thought to be involved in the pathogenesis of the ocular albinism with late-onset sensorineural deafness phenotype. Four transcript variants encoding two different isoforms have been found for this gene. This gene is highly similar to the Y chromosome TBL1Y gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBL1X/TBL1XR1
Polyclonal
Unconjugated
TBL1X
5330429M20; 8030499H02Rik; A630076E03Rik; AW539987; C21; C230089I12Rik; DC42; EBI; F-box-like/WD repeat protein TBL1XR1; F-box-like/WD repeat-containing protein TBL1X; F-box-like/WD repeat-containing protein TBL1XR1; Ira1; MRD41; nuclear receptor co-repressor/HDAC3 complex subunit; nuclear receptor corepressor/HDAC3 complex subunit TBLR1; SMAP55; TBL1; TBL1-related protein 1; Tbl1x; Tbl1xr1; TBLR1; transducin (beta)-like 1 X-linked; transducin (beta)-like 1 X-linked receptor 1; transducin (beta)-like 1X-linked; transducin (beta)-like 1X-linked receptor 1; transducin beta-like 1X-related protein 1; Transducin beta-like protein 1X; transducin-beta-like protein 1, X-linked
Goat
Antigen affinity chromatography
RUO
6907, 79718
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O60907, Q9BZK7
TBL1X, Tbl1xr1
Peptide with sequence SASDGSVCVLDLRK, from the C Terminus.
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.