Learn More
TBL1XR1 Monoclonal Antibody (OTI2A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811698S
This item is not returnable.
View return policy
Description
TBL1XR1 Monoclonal antibody specifically detects TBL1XR1 in Human samples. It is validated for Western BlotSpecifications
| TBL1XR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99618 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 83461 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2A8 | |
| Unconjugated | |
| CDCA3 | |
| 8030499H02Rik; A630076E03Rik; AW539987; C21; C230089I12Rik; DC42; F-box-like/WD repeat protein TBL1XR1; F-box-like/WD repeat-containing protein TBL1XR1; FLJ12894; Ira1; MRD41; nuclear receptor co-repressor/HDAC3 complex subunit; nuclear receptor corepressor/HDAC3 complex subunit TBLR1; RGD1560053; TBL1-related protein 1; TBL1XR1; TBLR1; transducin (beta)-like 1 X-linked receptor 1; transducin (beta)-like 1X-linked receptor 1; transducin beta-like 1X-related protein 1 | |
| Human recombinant protein fragment corresponding to amino acids 152-252 of human CDCA3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |