missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBL1Y Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139812
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QASAAATEAS AMAKAATMTP AAISQQ Highest antigen sequence indentity to the following orthologs: Rat - 58%, Mouse - 58%.
The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation. This encoded protein is found as a subunit in corepressor SMRT (silencing mediator for retinoid and thyroid receptors) complex along with histone deacetylase 3 protein. This gene is located adjacent to the ocular albinism gene and it is thought to be involved in the pathogenesis of the ocular albinism with late-onset sensorineural deafness phenotype. Four transcript variants encoding two different isoforms have been found for this gene. This gene is highly similar to the Y chromosome TBL1Y gene.
Specifications
| TBL1Y | |
| Polyclonal | |
| Unconjugated | |
| TBL1Y | |
| F-box-like/WD repeat-containing protein TBL1Y; TBL1; TBL1Y; transducin (beta)-like 1, Y-linked; transducin beta-like 1; transducin beta-like protein 1Y; Transducin-beta-like protein 1, Y-linked | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| Lot-Specific | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BQ87 | |
| TBL1Y | |
| Recombinant protein corresponding to Human TBL1Y. Recombinant protein control fragment (Product #RP-99599). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction