missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBL3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94540-0.1ml
This item is not returnable.
View return policy
Description
TBL3 Polyclonal antibody specifically detects TBL3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TBL3 | |
| Polyclonal | |
| Western Blot 1:1000-1:4000 | |
| SAZDWD repeat-containing protein SAZD, transducin (beta)-like 3, transducin beta-like protein 3, WD-repeat protein SAZD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 579-808 of human TBL3 (NP_006444.2). LSSGSDGLVKLWTIKNNECVRTLDAHEDKVWGLHCSRLDDHALTGASDSRVILWKDVTEAEQAEEQARQEEQVVRQQELDNLLHEKRYLRALGLAISLDRPHTVLTVIQAIRRDPEACEKLEATMLRLRRDQKEALLRFCVTWNTNSRHCHEAQAVLGVLLRREAPEELLAYEGVRAALEALLPYTERHFQRLSRTLQAAAFLDFLWHNMKLPVPAAAPTPWETHKGALP | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 10607 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction