missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBL3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97803H
This item is not returnable.
View return policy
Description
TBL3 Polyclonal antibody specifically detects TBL3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| TBL3 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| SAZDWD repeat-containing protein SAZD, transducin (beta)-like 3, transducin beta-like protein 3, WD-repeat protein SAZD | |
| Produced in rabbits immunized with E. coli-derived Human TBL3 fragment. | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 10607 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction