missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBPL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563273
This item is not returnable.
View return policy
Description
Immunogen sequence: EVTQENKDQP VISKHETEEN SESQSPQSRL PSPSEQDVGL GLNSSSLSNS HSQLHPGDTD SVQPSPEKPN SDS Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 47%.
TBPL2 is a transcription factor required in complex with TAF3 for the differentiation of myoblasts into myocytes. The complex replaces TFIID at specific promoters at an early stage in the differentiation process.
Specifications
| TBPL2 | |
| Polyclonal | |
| Unconjugated | |
| TBPL2 | |
| Gm348; TATA box binding protein like 2; TATA box binding protein-like 2; TATA box-binding protein-like protein 2; TATA box-binding protein-related factor 3; TATA-box binding protein like 2; TBP2; TBPL2; tbpl2 {ECO:0000250; TBP-like protein 2; TBP-related factor 3; TBP-related factor 3 (Trf3); TRF3; UniProtKB:P62380} | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 387332 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6SJ96 | |
| TBPL2 | |
| Recombinant protein corresponding to Human TBPL2. Recombinant protein control fragment (Product #RP-99955). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction