Learn More
TBR1 Antibody (2E10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26291-50ul
 View more versions of this product
Product Code. 30029190
  • € 343.00 / 50µL
Description
TBR1 Monoclonal antibody specifically detects TBR1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TBR1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA
2E10
ELISA
MGC141978, T-box brain protein 1, T-box, brain, 1, TBR-1, T-brain-1, TES-56
A synthesized peptide derived from Human TBR1 [UniProt Q16650]
50 μL
Neuroscience, Signal Transduction
10716
Store at -20 to -70C. Avoid freeze-thaw cycles.