missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBRG4 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98536JF646
This item is not returnable.
View return policy
Description
TBRG4 Polyclonal specifically detects TBRG4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TBRG4 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| cell cycle progression 2 protein, Cell cycle progression protein 2, Cell cycle progression restoration protein 2, CPR2, FAST kinase domains 4, FASTKD4Cpr2, KIAA0948H_TD2522F11.8, protein TBRG4, transforming growth factor beta regulator 4FAST kinase domain-containing protein 4 | |
| Produced in rabbits immunized with E. coli-derived Human TBRG4 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 9238 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction