missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBRG4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35595-20ul
This item is not returnable.
View return policy
Description
TBRG4 Polyclonal antibody specifically detects TBRG4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| TBRG4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| cell cycle progression 2 protein, Cell cycle progression protein 2, Cell cycle progression restoration protein 2, CPR2, FAST kinase domains 4, FASTKD4Cpr2, KIAA0948H_TD2522F11.8, protein TBRG4, transforming growth factor beta regulator 4FAST kinase domain-containing protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 382-631 of human TBRG4 (NP_004740.2).,, Sequence:, HPDQEDQFFSLVHEKLGSELPGLEPALQVDLVWALCVLQQAREAELQAVLHPEFHIQFLGGKSQKDQNTFQKLLHINATALLEYPEYSGPLLPASAVAPGPSALDRKVTPLQKELQETLKGLLGSADKGSLEVATQYGWVLDAEVLLDSDGEFLPVRDFVAPHLAQPTGSQSPPPGSKRLAFLRWEFPNFNSRSKDLLGRFVLARRHIVAAGFLIVDVPFYEWLELKSEWQKGAYLKDKMRKAVAEELAK | |
| 20 μL | |
| Cell Cycle and Replication | |
| 9238 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction