Learn More
TBX19 (Tpit) Monoclonal Antibody (OTI1C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800675S
Encompass_Preferred
 View more versions of this product
Product Code. 17578752
  • € 242.00 / 30µL
Description
TBX19 (Tpit) Monoclonal antibody specifically detects TBX19 (Tpit) in Human samples. It is validated for Western Blot
Specifications
Specifications
TBX19 (Tpit)
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P06729
Mouse
Affinity chromatography
RUO
914
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1C9
Unconjugated
CD2
AU019250; D1Ertd754e; dJ747L4.1; si:ch211-155d24.2; T-box 19; T-box factor, pituitary; T-box protein 19; T-box transcription factor 19; T-box transcription factor TBX19; T-box transcription factor TBX19; T-box-containing protein TBXT; TBS 19; TBS19; TBX19; tbx-19; Tpit; Tpit; Tbx19
Full length human recombinant protein of human CD2 produced in HEK293T cell.
30 μL