Learn More
TBX19 (Tpit) Monoclonal Antibody (OTI2G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800683
This item is not returnable.
View return policy
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Mutations in this gene were found in patients with isolated deficiency of pituitary POMC-derived ACTH, suggesting an essential role for this gene in differentiation of the pituitary POMC lineage. ACTH deficiency is characterized by adrenal insufficiency symptoms such as weight loss, lack of appetite (anorexia), weakness, nausea, vomiting, and low blood pressure.Specifications
| TBX19 (Tpit) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60806 | |
| TBX19 | |
| Human recombit protein fragment corresponding to amino acids 1-238 of human TBX19 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2G1 | |
| Unconjugated | |
| TBX19 | |
| TBS19, TPIT, dJ747L4.1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9095 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |