Learn More
TBX19 (Tpit) Monoclonal Antibody (OTI2G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800683S
This item is not returnable.
View return policy
Description
TBX19 (Tpit) Monoclonal antibody specifically detects TBX19 (Tpit) in Human samples. It is validated for Western BlotSpecifications
| TBX19 (Tpit) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IUC6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 148022 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G1 | |
| Unconjugated | |
| TICAM1 | |
| AU019250; D1Ertd754e; dJ747L4.1; si:ch211-155d24.2; T-box 19; T-box factor, pituitary; T-box protein 19; T-box transcription factor 19; T-box transcription factor TBX19; T-box transcription factor TBX19; T-box-containing protein TBXT; TBS 19; TBS19; TBX19; tbx-19; Tpit; Tpit; Tbx19 | |
| Human recombinant protein fragment corresponding to amino acids 120-420 of human TICAM1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |