missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX20 Monoclonal Antibody (OTI3D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809355S
This item is not returnable.
View return policy
Description
TBX20 Monoclonal antibody specifically detects TBX20 in Human samples. It is validated for Western BlotSpecifications
| TBX20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48739 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23760 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3D12 | |
| Unconjugated | |
| PITPNB | |
| 9430010M06Rik; AL022859; ASD4; cb826; H15-related T-box transcription factor hrT; hrt; T-box 20; T-box protein 20; T-box transcription factor; T-box transcription factor 20; T-box transcription factor TBX20; T-box transcription factor TBX20-like; Tbx12; TBX20; whz; ZF-TBX20 | |
| Full length human recombinant protein of human PITPNB produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction