Learn More
TBX20 Mouse anti-Human, Clone: OTI5D4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809303
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TBX20 is a member of the T-box transcription factor family expressed in the developing heart, eye, ventral neural tube, and limbs, indicating a possible role in regulating development of these tissues.Specifications
| TBX20 | |
| Monoclonal | |
| Unconjugated | |
| TBX20 | |
| ASD4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57057 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI5D4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UMR3 | |
| TBX20 | |
| Human recombit protein fragment corresponding to amino acids 1-340 of human TBX20 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.