Learn More
TBX3 Monoclonal Antibody (OTI8G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811209S
This item is not returnable.
View return policy
Description
TBX3 Monoclonal antibody specifically detects TBX3 in Human samples. It is validated for Western BlotSpecifications
| TBX3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q02952 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9590 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8G5 | |
| Unconjugated | |
| AKAP12 | |
| bladder cancer related protein XHL; D5Ertd189e; T-box 3; T-box 3 (ulnar mammary syndrome); T-box protein 3; T-box transcription factor TBX3; TBX3; TBX3-ISO; UMS; XHL | |
| Human recombinant protein fragment corresponding to amino acids 266-557 of human AKAP12 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |