Learn More
TBX5 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99529PCP
This item is not returnable.
View return policy
Description
TBX5 Polyclonal antibody specifically detects TBX5 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Immunohistochemistry (Paraffin) | |
| PerCP |