missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBXA2R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118797
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TAS2R.
The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.
Specifications
| TBXA2R | |
| Polyclonal | |
| Unconjugated | |
| TBXA2R | |
| BDPLT13; prostanoid TP receptor; TBXA2R; Thromboxane A2 receptor; thromboxane prostanoid; Thromboxane receptor; TP; TXA2 receptor; TXA2-R; TXR2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21390, 24816, 6915 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P21731, P30987, P34978 | |
| TBXA2R | |
| A synthesized peptide derived from human TBXA2R(Accession P21731), corresponding to amino acid residues V263-R313. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction