missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBXAS Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501364S
This item is not returnable.
View return policy
Description
TBXAS Monoclonal antibody specifically detects TBXAS in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| TBXAS | |
| Monoclonal | |
| 0.94 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7L9L4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 92597 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Western Blot | |
| OTI1B8 | |
| Unconjugated | |
| MOB1B | |
| BDPLT14; CYP5; Cyp5a1; cytochrome P450 5A1; cytochrome P450, family 5, subfamily A, polypeptide 1; FLJ52771; GHOSAL; platelet, cytochrome P450, subfamily V; similar to Thromboxane-A synthase (TXA synthase) (TXS); Tbxas1; THAS; ThromboxA ane synthase 1; thromboxane A synthase 1; thromboxane A synthase 1 (platelet); thromboxane A synthase 1 (platelet, cytochrome P450, family 5, subfamily A); thromboxane A synthase 1, platelet; thromboxane synthase; thromboxane-A synthase; TS; TXA synthase; TXAS; TXS | |
| Full length human recombinant protein of human MOBKL1A produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction