missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-N 1752

Product Code. 18004242
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18004242

Brand: Tocris Bioscience™ 4435/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NaV channel blocker

TC-N 1752 is a human NaV channel inhibitor (IC50 values are 0.17, 0.3, 0.4, 1.1 and 1.6 μM at hNaV1.7, hNaV1.3, hNaV1.4, hNaV1.5 and hNav1.9 respectively). Also inhibits tetrodotoxin -sensitive sodium channels. Displays analgesic efficacy in the formalin pain model.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1211866-85-1
Molecular Formula C25H27F3N6O3

Specifications

Quantity 50 mg
Formula Weight 516.52
Purity >99%
Molecular Weight (g/mol) 516.525
Chemical Name or Material TC-N 1752
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.