Learn More
TCAP Monoclonal Antibody (OTI7H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808341S
Encompass_Preferred
 View more versions of this product
Product Code. 17549493
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
TCAP Monoclonal antibody specifically detects TCAP in Human samples. It is validated for Western Blot
Specifications
Specifications
TCAP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O15273
Mouse
Affinity chromatography
RUO
8557
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI7H5
Unconjugated
Tcap
19 kDa sarcomeric protein; AI503981; AJ223855; CMD1N; CMH25; LGMD2G; limb girdle muscular dystrophy 2G (autosomal recessive); Tcap; T-cap; TELE; telethonin; teneurin C-terminal associated peptide; titin cap protein; titin-cap; titin-cap (telethonin)
Full length human recombinant protein of human TCAP produced in E.coli.
30 μL