Learn More
Tocris Bioscience™ TCB-2
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TCB-2 is a high affinity 5-HT2A receptor agonist (Ki values are 0.73 and 0.75 nM for rat and human receptors respectively). Potently stimulates IP3 accumulation in NIH3T3 cells stably expressing rat 5-HT2A receptors (EC50 = 36 nM). Induces head twitches and hypothermia in mice following i.p. administration.
Specifications
Specifications
| Chemical Name or Material | (4-Bromo-3,6-dimethoxybenzocyclobuten-1-yl)methylamine hydrobromide |
| CAS | 912342-28-0 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C11H14BrNO2.HBr |
| Formula Weight | Observed MW: 353.05 |