missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCCR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5141064
This item is not returnable.
View return policy
Description
TCCR Polyclonal Antibody for Western Blot, Flow
In mice, CD4+ helper T-cells differentiate into type 1 (Th1) cells and type 2 (Th2) cells, which are critical for cell-mediated immunity and most antibody responses, respectively. The differentiation of Th1 cells is predominantly under the influence of IL12, while IL4 influences the differentiation of Th2 cells. Mice deficient in these cytokines, their receptors, or associated transcription factors have impaired, but not absent, Th1 or Th2 immune responses. The gene discussed here encodes a protein that is similar to the mouse T-cell cytokine receptor Tccr at the amino acid level and is predicted to be a glycosylated transmembrane protein. Diseases associated with IL27RA include Hyper Ige Recurrent Infection Syndrome 4 and Crisponi/Cold-Induced Sweating Syndrome 2.
Specifications
| TCCR | |
| Polyclonal | |
| Unconjugated | |
| Il27ra | |
| class I cytokine receptor; CRL1; CRL1IL-27R; cytokine receptor family, class 1 (WSXWS), member 1; cytokine receptor WSX-1; Cytokine receptor-like 1; il-27 receptor; IL-27 receptor subunit alpha; IL27R; IL-27R; IL-27R subunit alpha; IL27RA; IL-27RA; IL-27R-alpha; interleukin 27 receptor subunit alpha; interleukin 27 receptor, alpha; interleukin-27 receptor subunit alpha; T cell cytokine receptor; T cell cytokine receptor; cytokine receptor family, class 1 (WSXWS), member 1; TCCR; T-cell cytokine receptor type 1; Type I T-cell cytokine receptor; UNQ296/PRO336; WSX1; WSX-1; ZcytoR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9466 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6UWB1 | |
| Il27ra | |
| Synthetic peptide within human WSX-1 aa 71-120. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction