Learn More
TCEAL1 Mouse anti-Human, Clone: OTI2G10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807701
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TCEAL1 (transcription elongation factor A (SII) protein 3), also known as p21, SIIR or pp21, is a member of the TFS-II family. Transcription elongation factors of the TFS-II family are responsible for releasing RNA polymerase II (Pol II) from transcriptional arrest. DNA arresting sites can result in locked ternary complexes if elongating RNA polymerases are trapped. Transcription elongation factors function to activate the intrinsic RNA cleavage activity of RNA polymerases. This allows the RNA polymerase to cleave the nascent transcript, thereby forming a new 3′-terminus to resume elongation. TCEAL1 is a 157 amino acid nuclear protein that is ubiquitously expressed. TCEAL1 may exert its effects via protein-protein interactions with other transcriptional regulators rather than via direct binding of DNA.Specifications
| TCEAL1 | |
| Monoclonal | |
| Unconjugated | |
| TCEAL1 | |
| SIIR, WEX9, p21, pp21 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9338 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q15170 | |
| TCEAL1 | |
| Full length human recombit protein of human TCEAL1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.