missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEANC2 Monoclonal Antibody (OTI8E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809333
This item is not returnable.
View return policy
Description
TCEANC2 (Transcription Elongation Factor A N-Terminal And Central Domain Containing 2) is a Protein Coding gene.Specifications
| TCEANC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96MN5 | |
| TCEANC2 | |
| Full length human recombit protein of human C1orf83 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8E11 | |
| Unconjugated | |
| TCEANC2 | |
| C1orf83 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 127428 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction