missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEANC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561372
This item is not returnable.
View return policy
Description
Immunogen sequence: TMLELPDQTK ENLVEALQEL KKKIPSREVL KSTRIGHTVN KMRKHSDSEV ASLAREVYTE WKTFTEKHSN RPSIEV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%.
TCEANC2 (Transcription Elongation Factor A N-Terminal And Central Domain Containing 2) is a Protein Coding gene.
Specifications
| TCEANC2 | |
| Polyclonal | |
| Unconjugated | |
| Tceanc2 | |
| 2210010B22Rik; 2210012G02Rik; 6330404A07Rik; AU015593; C1orf83; TCEANC2; Tdeanc2; transcription elongation factor A (SII) N-terminal and central domain containing 2; transcription elongation factor A N-terminal and central domain containing 2; transcription elongation factor A N-terminal and central domain-containing protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127428 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MN5 | |
| Tceanc2 | |
| Recombinant protein corresponding to Human TCEANC2. Recombinant protein control fragment (Product #RP-94066). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction