missing translation for 'onlineSavingsMsg'
Learn More

TCEB3B Antibody [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30516336 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30516336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30516336 Supplier Novus Biologicals Supplier No. NBP338555V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCEB3B Polyclonal antibody specifically detects TCEB3B in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEB3B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias 110kDa Elongin A, EloA2, Elongin-A2, HsT832, MGC119351, RNA polymerase II transcription factor SIII subunit A2, TCEB3Ltranscription elongation factor (SIII) elongin A2 elongin A2, Transcription elongation factor B polypeptide 3B, transcription elongation factor B polypeptide 3B (elongin A2)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 534-753 of human TCEB3B (NP_057511.2).,, Sequence:, AQVLRNNPDALSDVGEVPYWVLEPVLEGWRPDQLYRRKKDNHALVRETDELRRNHCFQDFKEEKPQENKTWREQYLRLPDAPEQRLRVMTTNIRSARGNNPNGREAKMICFKSVAKTPYDTSRRQEKSAGDADPENGEIKPASKPAGSSHTPSSQSSSGGGRDSSSSILRWLPEKRANPCLSSSNEHAAPAAKTRKQAAKKVAPLMAKAIRDYKRRFSRR
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51224
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.