missing translation for 'onlineSavingsMsg'
Learn More

TCEB3B Antibody, Novus Biologicals™

Product Code. 18455101 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18455101 25 μL 25µL
18052577 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18455101 Supplier Novus Biologicals Supplier No. NBP19248225ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCEB3B Polyclonal specifically detects TCEB3B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEB3B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias 110kDa Elongin A, EloA2, Elongin-A2, HsT832, MGC119351, RNA polymerase II transcription factor SIII subunit A2, TCEB3Ltranscription elongation factor (SIII) elongin A2 elongin A2, Transcription elongation factor B polypeptide 3B, transcription elongation factor B polypeptide 3B (elongin A2)
Gene Symbols TCEB3B
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:GHKSSRQEKRPLCAQGDWHSPTLIREKSCGACLREETPRMPSWASARDRQPSDFKTDKEGGQAGSGQRVPALEEAPDSHQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51224
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.