missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-12/HTF4 Antibody (OTI4D6), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74473G
This item is not returnable.
View return policy
Description
TCF-12/HTF4 Monoclonal specifically detects TCF-12/HTF4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TCF-12/HTF4 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TCF12 | |
| Recombinant protein fragment corresponding to amino acids 1-250 of human TCF-12/HTF4 (NP_996919) produced in E.coli. | |
| 0.1 mL | |
| 6938.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4D6 | |
| Western Blot, Immunohistochemistry | |
| BHLHB20, bHLHb20E-box-binding protein, Class B basic helix-loop-helix protein 20, HEBDNA-binding protein HTF4, helix-loop-helix transcription factor 4, HsT17266, HTF4TCF-12, transcription factor 12, Transcription factor HTF-4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction