missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-2/HNF-1 beta Antibody (SR2073), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | TCF-2/HNF-1 beta |
|---|---|
| Clone | SR2073 |
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663744
|
Novus Biologicals
NBP3-21691-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680204
|
Novus Biologicals
NBP3-21691-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCF-2/HNF-1 beta Monoclonal antibody specifically detects TCF-2/HNF-1 beta in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TCF-2/HNF-1 beta | |
| Western Blot 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FJHN, hepatocyte nuclear factor 1-beta, HNF1 beta A, HNF1 homeobox B, HNF-1B, HNF1beta, HNF-1-beta, HNF2, Homeoprotein LFB3, LFB3, LF-B3, MODY5HPC11, TCF-2, TCF2transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor, Transcription factor 2, transcription factor 2, hepatic, Variant hepatic nuclear factor 1, vHNF1 | |
| A synthesized peptide derived from human TCF-2/HNF-1 beta (Uniprot #: P35680) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2073 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Diabetes Research | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6928 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title