Learn More
TCF12 Monoclonal Antibody (OTI1F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809700S
Encompass_Preferred
 View more versions of this product
Product Code. 17532204
  • € 242.00 / 30µL
Description
TCF12 Monoclonal antibody specifically detects TCF12 in Human samples. It is validated for Western Blot
Specifications
Specifications
TCF12
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96JM7
Mouse
Affinity chromatography
RUO
84456
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1F11
Unconjugated
L3MBTL3
A130037E08Rik; ALF1; bHLHb20; Class A helix-loop-helix transcription factor ME1; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E-box-binding protein; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; ME1; Meb; REB; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; TCF12; TCF-12; Transcription factor 12; transcription factor HTF-4
Human recombinant protein fragment corresponding to amino acids 98-319 of human L3MBTL3 produced in E.coli.