missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF12 Monoclonal Antibody (OTI3C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809697
This item is not returnable.
View return policy
Description
TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.Specifications
| TCF12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99081 | |
| TCF12 | |
| Full length human recombit protein of human TCF12 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C11 | |
| Unconjugated | |
| TCF12 | |
| CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6938 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction