missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF12 Monoclonal Antibody (OTI4D6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526671
This item is not returnable.
View return policy
Description
TCF12 Monoclonal Antibody for Western Blot, IHC (P)
TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.
Specifications
| TCF12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99081 | |
| TCF12 | |
| Human recombinant protein fragment corresponding to amino acids 1-250 of TCF12 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4D6 | |
| Unconjugated | |
| TCF12 | |
| A130037E08Rik; ALF1; bHLHb20; Class A helix-loop-helix transcription factor ME1; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E-box-binding protein; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; ME1; Meb; REB; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; TCF12; TCF-12; Transcription factor 12; transcription factor HTF-4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6938 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction