missing translation for 'onlineSavingsMsg'
Learn More

TCF12 Mouse anti-Human, Clone: OTI4D6, liquid, TrueMAB™

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA804474

Encompass_Preferred

Product Code. 17594063

  • € 540.87 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.
Specifications

Specifications

TCF12
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q99081
TCF12
Human recombit protein fragment corresponding to amino acids 1-250 of human TCF12 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI4D6
Unconjugated
TCF12
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
Mouse
Affinity Chromatography
RUO
6938
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.