Learn More
TCF3 Rabbit anti-Human, Polyclonal , Abnova™
TCF3 polyclonal antibody raised against recombinant human TCF3.
Brand: Abnova PAB29500.100uL
Description
The TCF3 gene, also called E2A, encodes 2 basic helix-loop-helix (bHLH) transcription factors, E12 and E47, through alternative splicing. E12 and E47 are involved in regulation of immunoglobulin gene expression (Bain et al., 1994 [PubMed 8001125]).[supplied by OMIM]
Sequence: RTFSEGTHFTESHSSLSSSTFLGPGLGGKSGERGAYASFGRDAGVGGLTQAGFLSGELALNSPGPLSPSGMKGTSQYYPSSpecifications
| TCF3 | |
| Polyclonal | |
| TCF3 polyclonal antibody raised against recombinant human TCF3. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| TCF3 | |
| Recombinant protein corresponding to amino acids of human TCF3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| E2A/ITF1/MGC129647/MGC129648/bHLHb21 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6929 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.