missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF4 Antibody (SR2168), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 472.00
Specifications
| Antigen | TCF4 |
|---|---|
| Clone | SR2168 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:80, Immunohistochemistry 1:50-1:200, Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, ChIP Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668884
|
Novus Biologicals
NBP3-22305-100ul |
100 μg |
€ 472.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641214
|
Novus Biologicals
NBP3-22305-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ChIP Assay (ChIP)Specifications
| TCF4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:80, Immunohistochemistry 1:50-1:200, Chromatin Immunoprecipitation (ChIP) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4 | |
| A synthesized peptide derived from human TCF4 (Uniprot #: P15884) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2168 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, ChIP Assay | |
| Unconjugated | |
| Rabbit | |
| Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6925 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title